Skip to Content
Merck

AABH9BD8A681

AA BLOCKS, INC.

6,7-Dimethoxy-3,4-dihydroisoquinoline-2(1H)-carboximidamide sulfate

Synonym(s):

sulfuric acid compound with 6,7-dimethoxy-3,4-dihydro-2(1H)-isoquinolinecarboximidamide (1:1)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H19O6N3S1
CAS Number:
Molecular Weight:
333.37
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H17N3O2.H2O4S/c1-16-10-5-8-3-4-15(12(13)14)7-9(8)6-11(10)17-2;1-5(2,3)4/h5-6H,3-4,7H2,1-2H3,(H3,13,14);(H2,1,2,3,4)

Inchi Key

LDGBVLZTLHOPBA-UHFFFAOYSA-N

Storage temp.

N/A

Country of Origin

US





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library