Skip to Content
Merck

COM448668429

Combi-Blocks, Inc.

Diphenylammonium trifluoromethanesulfonate

Synonym(s):

trifluoromethanesulfonic acid compound with N,N-diphenylamine (1:1)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C13H12O3N1S1F3
CAS Number:
Molecular Weight:
319.3
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H11N.CHF3O3S/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;2-1(3,4)8(5,6)7/h1-10,13H;(H,5,6,7)

Inchi Key

MGEGQAUINMTPGX-UHFFFAOYSA-N

Purity

98%

Country of Origin

CN



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library