Skip to Content
Merck

BDPH9BCC99A5

Bide Pharmatech

1,3,5,7,9,11,13,15-Octaphenyl-2,4,6,8,10,12,14,16,17,18,19,20-dodecaoxa-1,3,5,7,9,11,13,15-octasilapentacyclo[9.5.1.13,9.15,15.17,13]icosane

Synonym(s):

1,3,5,7,9,11,13,15-octaphenylpentacyclo[9.5.1.1~3,9~.1~5,15~.1~7,13~]octasiloxane

Select a Size

Change View

About This Item

Linear Formula:
C48H40O12Si8
CAS Number:
Molecular Weight:
1033.52
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C48H40O12Si8/c1-9-25-41(26-10-1)61-49-62(42-27-11-2-12-28-42)52-65(45-33-17-5-18-34-45)54-63(50-61,43-29-13-3-14-30-43)56-67(47-37-21-7-22-38-47)57-64(51-61,44-31-15-4-16-32-44)55-66(53-62,46-35-19-6-20-36-46)59-68(58-65,60-67)48-39-23-8-24-40-48/h1-40H

Inchi Key

KBXJHRABGYYAFC-UHFFFAOYSA-N

Shipping Temperature

Cold-chain transportation

Purity

98%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Form: Crystal - Powder / Colour: White - Almost white




Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library