Skip to Content
Merck

BDPH9BCCF3CF

Bide Pharmatech

3-Bromo-5-fluorobenzene-1-sulfonyl chloride

Synonym(s):

3-bromo-5-fluorobenzenesulfonyl chloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H3O2Cl1Br1S1F1
CAS Number:
Molecular Weight:
273.51
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H3BrClFO2S/c7-4-1-5(9)3-6(2-4)12(8,10)11/h1-3H

Inchi Key

HZJBQCUMWMMHMK-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Colorless to Yellow to Yellow-brown Liquid



Still not finding the right product?


hstatements

Precautionary Statements

Signal Word

Danger

Pictograms

Corrosion


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library