Merck
Proanthocyanidins rapidly stabilize the demineralized dentin layer.
Journal of dental research (2013-06-01)
Y Liu, V Dusevich, Y Wang
ABSTRACT
While proanthocyanidins (PA) are effective in improving collagen's resistance to collagenolytic degradation, the direct incorporation of PA into an adhesive system is detrimental to the light-curing thereof. Conversely, the use of PA as a primer could circumvent this issue, but little is known about the efficacy of PA in stabilizing collagen when applied in a clinically relevant manner. This study investigated the pre- and post-digestion morphology of an acid-etched dentin collagen layer that underwent PA treatment for time periods on a scale of seconds. The null hypothesis, that there is no difference between the PA-treated and untreated control group, had to be rejected, since it was revealed that the untreated control could not survive 1 hr of exogenous collagenase digestion, while the PA-treated collagen could sustain at least 16 hrs of digestion with no perceptible changes in collagen structure. In addition, the stabilizing effect of the gold-standard cross-linker glutaraldehyde at comparable experimental conditions was found to be almost non-existent within the 5, 15, or 30 sec of cross-linking permitted. Therefore, PA have been proven to be extraordinarily efficient in stabilizing demineralized dentin collagen against enzymatic challenges in a clinically relevant setting, likely due to the non-covalent nature of their interaction with collagen molecules.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Silicon carbide, -200 mesh particle size
Sigma-Aldrich
Silicon carbide, nanopowder, <100 nm particle size
Sigma-Aldrich
Silicon carbide, −400 mesh particle size, ≥97.5%
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, puriss. p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Phosphoric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, 85.0-88.0%
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis