Merck
  • MiR-1298 affects cell proliferation and apoptosis in C6 cells by targeting SET domain containing 7.
MiR-1298 affects cell proliferation and apoptosis in C6 cells by targeting SET domain containing 7.
International journal of immunopathology and pharmacology (2017-08-02)
Chun-Mei Wang, Bao-Hua Cheng, Qing-Jie Xue, Jing Chen, Bo Bai
ABSTRACT
Our previous high-throughput sequencing indicated that rno-miR-1298 was down-regulated in ischemia-reperfusion model of rat. However, little is known about the function and molecular mechanism of rno-miR-1298 in rat tumor cell. In this study, rno-miR-1298 was detected to be significantly down-regulated in rat tumor C6 cells. Moreover, overexpression of rno-miR-1298 obviously inhibited the proliferation and induced apoptosis in C6 cells. SET domain containing 7 (SETD 7) was identified to be a target of rno-miR-1298 using bioinformatics and luciferase reporter assays. Overexpression of rno-miR-1298 markedly reduced the expression of SETD 7 at protein level. Knockdown of SETD 7 also suppressed proliferation and promoted apoptosis in C6 cells. It was indicated that rno-miR-1298 affected cell proliferation and apoptosis of rat tumor cells by targeting SETD 7. Thus, the newly identified miR-1298/SETD 7 expands the elaboration of the mechanisms of the development and progression of tumors and may provide therapeutic target for tumors of nervous system.