Skip to Content

BDPH9BCDBD0A

Bide Pharmatech

4-(1-(o-Tolyl)vinyl)benzoic acid

Synonym(s):

4-(1-(o-tolyl)vinyl)benzoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C16H14O2
CAS Number:
Molecular Weight:
238.29
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C16H14O2/c1-11-5-3-4-6-15(11)12(2)13-7-9-14(10-8-13)16(17)18/h3-10H,2H2,1H3,(H,17,18)

Inchi Key

KMUWACOLXILRKG-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 4-(1-(o-Tolyl)vinyl)benzoic acid


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library