Skip to Content
Merck

AABH93DE4E5D

AA BLOCKS, INC.

7-APB (hydrochloride)

Synonym(s):

1-(benzofuran-7-yl)propan-2-amine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H14O1N1Cl1
CAS Number:
Molecular Weight:
211.69
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C11H13NO.ClH/c1-8(12)7-10-4-2-3-9-5-6-13-11(9)10;/h2-6,8H,7,12H2,1H3;1H

Inchi Key

OENDNRHBWDLDMM-UHFFFAOYSA-N

Purity

98%

Storage temp.

N/A

Country of Origin

US





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library