Skip to Content
Merck

BDPH9BCC29A9

Bide Pharmatech

1-(2-Bromoethyl)-3-phenoxy-benzene

Synonym(s):

1-(2-bromoethyl)-3-phenoxybenzene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C14H13O1Br1
CAS Number:
Molecular Weight:
277.16
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Key

AGZTWRYOPSZJII-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Liquid

Inchi Code

1S/C14H13BrO/c15-10-9-12-5-4-8-14(11-12)16-13-6-2-1-3-7-13/h1-8,11H,9-10H2



Still not finding the right product?

Explore all of our products under 1-(2-Bromoethyl)-3-phenoxy-benzene



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library