Skip to Content
Merck

BDPH9BCCB85D

Bide Pharmatech

1-(3,4-Dichlorophenyl)ethan-1-amine hydrochloride

Synonym(s):

1-(3,4-dichlorophenyl)ethanamine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H10N1Cl3
CAS Number:
Molecular Weight:
226.53
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid

Inchi Code

1S/C8H9Cl2N.ClH/c1-5(11)6-2-3-7(9)8(10)4-6;/h2-5H,11H2,1H3;1H

Inchi Key

LTTRKULQEVDRNO-UHFFFAOYSA-N



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library