Skip to Content
Merck

BDPH9BCCF032

Bide Pharmatech

2,6-Dichloro-3-hydroxybenzonitrile

Synonym(s):

2,6-dichloro-3-hydroxybenzonitrile

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H3O1N1Cl2
CAS Number:
Molecular Weight:
188.01
MDL number:
Product BDPH9BCCF032 is not currently sold in your country.


Inchi Code

1S/C7H3Cl2NO/c8-5-1-2-6(11)7(9)4(5)3-10/h1-2,11H

Inchi Key

JAIIUXZFGCEYAH-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

96%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 2,6-Dichloro-3-hydroxybenzonitrile



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library