Skip to Content
Merck

BDPH9BCCF1CC

Bide Pharmatech

2-Amino-1H-imidazole-5-carboxylic acid

Synonym(s):

2-amino-1H-imidazole-5-carboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C4H5O2N3
CAS Number:
Molecular Weight:
127.1
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C4H5N3O2/c5-4-6-1-2(7-4)3(8)9/h1H,(H,8,9)(H3,5,6,7)

Inchi Key

YTYZMZQDGZMTJM-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library