Skip to Content
Merck

BDPH9BCD01DF

Bide Pharmatech

5-Bromo-2,3-dimethylpyrazine

Synonym(s):

5-bromo-2,3-dimethylpyrazine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H7N2Br1
CAS Number:
Molecular Weight:
187.04
MDL number:
Product BDPH9BCD01DF is not currently sold in your country.


Inchi Code

1S/C6H7BrN2/c1-4-5(2)9-6(7)3-8-4/h3H,1-2H3

Inchi Key

LACACUPDGFABKK-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Liquid or Solid or Semi-solid or lump



Still not finding the right product?

Explore all of our products under 5-Bromo-2,3-dimethylpyrazine


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library