Skip to Content
Merck

BDPH9BCD475C

Bide Pharmatech

2-Chloro-3,6-difluorobenzonitrile

Synonym(s):

2-chloro-3,6-difluorobenzonitrile

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H2N1Cl1F2
CAS Number:
Molecular Weight:
173.55
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H2ClF2N/c8-7-4(3-11)5(9)1-2-6(7)10/h1-2H

Inchi Key

AREWFYUZFHLWNH-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Colorless to yellow solid or liquid



Still not finding the right product?

Explore all of our products under 2-Chloro-3,6-difluorobenzonitrile




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library