Skip to Content
Merck

BDPH9BCD687D

Bide Pharmatech

(4-Methoxypyridin-3-yl)boronic acid hydrochloride

Synonym(s):

4-methoxy-3-pyridinylboronic acid hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H9O3N1Cl1B1
CAS Number:
Molecular Weight:
189.41
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H8BNO3.ClH/c1-11-6-2-3-8-4-5(6)7(9)10;/h2-4,9-10H,1H3;1H

Inchi Key

JLONCHBPEIKTEA-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Country of Origin

CN

Physical Form

Solid

Storage temp.

Inert atmosphere, 2-8°C



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library