Skip to Content
Merck

BDPH9BCD6EA7

Bide Pharmatech

3-Hydroxy-[1,1'-biphenyl]-4-carbonitrile

Synonym(s):

3-hydroxy[1,1'-biphenyl]-4-carbonitrile

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C13H9O1N1
CAS Number:
Molecular Weight:
195.22
MDL number:
Product BDPH9BCD6EA7 is not currently sold in your country.


Inchi Code

1S/C13H9NO/c14-9-12-7-6-11(8-13(12)15)10-4-2-1-3-5-10/h1-8,15H

Inchi Key

DKLJNQAGZBVPLO-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library