Skip to Content
Merck

Skip To

BDPH9BCDB5D2

Bide Pharmatech

N-(2-Cyanoethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

Synonym(s):

N-(2-cyanoethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C16H21O3N2B1
CAS Number:
Molecular Weight:
300.17
MDL number:
Product BDPH9BCDB5D2 is not currently sold in your country.


Inchi Code

1S/C16H21BN2O3/c1-15(2)16(3,4)22-17(21-15)13-8-6-12(7-9-13)14(20)19-11-5-10-18/h6-9H,5,11H2,1-4H3,(H,19,20)

Inchi Key

UKHWUDSILSYFKO-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library