Skip to Content
Merck

BDPH9BCDC7BB

Bide Pharmatech

1-(p-Tolyl)pyrrolidin-2-one

Synonym(s):

1-(4-methylphenyl)-2-pyrrolidinone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H13O1N1
CAS Number:
Molecular Weight:
175.23
MDL number:
Product BDPH9BCDC7BB is not currently sold in your country.


Inchi Code

1S/C11H13NO/c1-9-4-6-10(7-5-9)12-8-2-3-11(12)13/h4-7H,2-3,8H2,1H3

Inchi Key

ORPHOKOVVUFNKE-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?

Explore all of our products under 1-(p-Tolyl)pyrrolidin-2-one


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library