Skip to Content
Merck

BDPH9BCDCCF3

Bide Pharmatech

6-Methyl-1H-indazole-5-carbaldehyde

Synonym(s):

6-methyl-1H-indazole-5-carbaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H8O1N2
CAS Number:
Molecular Weight:
160.18
MDL number:
Product BDPH9BCDCCF3 is not currently sold in your country.


Inchi Code

1S/C9H8N2O/c1-6-2-9-7(4-10-11-9)3-8(6)5-12/h2-5H,1H3,(H,10,11)

Inchi Key

NKCNHLIMIJOJOM-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library