Skip to Content
Merck

BDPH9BCDF06E

Bide Pharmatech

2-Fluorobutan-1-amine hydrochloride

Synonym(s):

2-fluoro-1-butanamine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C4H11N1Cl1F1
CAS Number:
Molecular Weight:
127.59
MDL number:
Product BDPH9BCDF06E is not currently sold in your country.


Inchi Code

1S/C4H10FN.ClH/c1-2-4(5)3-6;/h4H,2-3,6H2,1H3;1H

Inchi Key

GCFJSNTVZBKQCG-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library