Skip to Content
Merck

BDPH9BCDFEB2

Bide Pharmatech

4-(1,1-Difluoroethyl)benzoic acid

Synonym(s):

4-(1,1-difluoroethyl)benzoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H8O2F2
CAS Number:
Molecular Weight:
186.16
MDL number:
Product BDPH9BCDFEB2 is not currently sold in your country.


Inchi Code

1S/C9H8F2O2/c1-9(10,11)7-4-2-6(3-5-7)8(12)13/h2-5H,1H3,(H,12,13)

Inchi Key

JYOQPEQDJAOTAU-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid or liquid



Still not finding the right product?

Explore all of our products under 4-(1,1-Difluoroethyl)benzoic acid


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library