Skip to Content
Merck

BDPH9BCE112C

Bide Pharmatech

2-(2-Methylbenzyl)succinic acid

Synonym(s):

2-(2-methylbenzyl)succinic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H14O4
CAS Number:
Molecular Weight:
222.24
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H14O4/c1-8-4-2-3-5-9(8)6-10(12(15)16)7-11(13)14/h2-5,10H,6-7H2,1H3,(H,13,14)(H,15,16)

Inchi Key

WFWVINOHIVDSEV-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 2-(2-Methylbenzyl)succinic acid



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library