Skip to Content
Merck

BDPH9C01E853

Bide Pharmatech

(S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-mercapto-1-oxopropan-2-yl)amino)-5-oxopentanoic acid

Synonym(s):

(2S)-2-amino-5-{[(1R)-2-[(carboxymethyl)amino]-2-oxo-1-(sulfanylmethyl)ethyl]amino}-5-oxopentanoic acid

Select a Size

Change View

About This Item

Linear Formula:
C10H17O6N3S1
CAS Number:
Molecular Weight:
307.33
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H17N3O6S/c11-5(10(18)19)1-2-7(14)13-6(4-20)9(17)12-3-8(15)16/h5-6,20H,1-4,11H2,(H,12,17)(H,13,14)(H,15,16)(H,18,19)/t5-,6-/m0/s1

Inchi Key

RWSXRVCMGQZWBV-WDSKDSINSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Form: Crystal - Powder / Colour: White





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library