Skip to Content
Merck

BDPH9C01EAEB

Bide Pharmatech

(1S,2S)-Cyclopropane-1,2-dicarboxylic acid

Synonym(s):

(1S,2S)-1,2-cyclopropanedicarboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H6O4
CAS Number:
Molecular Weight:
130.1
MDL number:
Product BDPH9C01EAEB is not currently sold in your country.


Storage temp.

Sealed in dry, 2-8°C

Inchi Code

1S/C5H6O4/c6-4(7)2-1-3(2)5(8)9/h2-3H,1H2,(H,6,7)(H,8,9)/t2-,3-/m0/s1

Inchi Key

RLWFMZKPPHHHCB-HRFVKAFMSA-N

Shipping Temperature

Normal

Purity

95%

Country of Origin

CN

Physical Form

White to yellow solid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library