Skip to Content
Merck

ST9H9BC1663E

Shanghai Topscience Co., Ltd.

CSF1R-IN-3

Synonym(s):

N-(5-(((3E)-6-((((5-(tert-butyl)isoxazol-3(2H)-ylidene)amino)(hydroxy)methylene)amino)-2-hydroxy-3H-indol-3-ylidene)methyl)-2,4-dimethyl-1H-pyrrol-3-yl)-2-(4-methylpiperazin-1-yl)acetimidic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C30H38N8O4/c1-17-22(31-18(2)27(17)35-26(39)16-38-11-9-37(6)10-12-38)14-21-20-8-7-19(13-23(20)33-28(21)40)32-29(41)34-25-15-24(42-36-25)30(3,4)5/h7-8,13-15,31H,9-12,16H2,1-6H3,(H,33,40)(H,35,39)(H2,32,34,36,41)/b21-14-

Inchi Key

YLKFFTVAKILRBN-STZFKDTASA-N

Shipping Temperature

Shipping with blue ice/Shipping at ambient temperature.

Purity

98.40%

Storage temp.

Powder: -20C for 3 years | In solvent: -80C for 1 year@Shipping with blue ice/Shipping at ambient temperature.

Country of Origin

CN



Still not finding the right product?

Explore all of our products under CSF1R-IN-3




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library