Skip to Content
Merck
Iodine solution c(I2) = 0.05 mol/l (0.1 N), Titripur®, reag. Ph. Eur.

Iodine solution

Iodine solution c(I2) = 0.05 mol/l (0.1 N), Titripur®, reag. Ph. Eur.

Products

Product Category
Brand
Physical Form
Product Line
Quality Segment
Technique
Showing 1-3 of 3