Merck
  • Molecular cloning of the small (gamma) subunit of human TFIIA reveals functions critical for activated transcription.
Molecular cloning of the small (gamma) subunit of human TFIIA reveals functions critical for activated transcription.
Genes & development (1994-10-01)
J Ozer, P A Moore, A H Bolden, A Lee, C A Rosen, P M Lieberman
ABSTRACT
TFIIA is thought to play an important role in transcriptional regulation in higher eukaryotes, but its precise function is unclear. A human cDNA encoding a protein with 45% identity to the small subunit of yeast TFIIA has been isolated. TFIIA activity could be reconstituted by the mixing of recombinant large (alpha beta) and small (gamma) subunits. TFIIA-depleted HeLa nuclear extracts were used to demonstrate that TFIIA is essential for basal and activated transcription by several distinct classes of activators. Recombinant TFIIA functioned in transcriptional activation whether expressed as a dimer (alpha beta+gamma) or as a trimer (alpha+beta+gamma), which closely resembles the native form. Yeast TFIIA also functioned in transcriptional activation, and the human gamma subunit was functionally interchangeable with TOA2, its yeast homolog. Recombinant TFIIA mediated the stimulation of TFIID binding to the TATA region and downstream promoter sequences by the Zta transcriptional activator. Significantly, we found that TFIIA bound directly to Zta in an activation domain-dependent manner. One consequence of the TFIIA-mediated interaction between Zta and TFIID was the formation of a promoter-bound complex resistant to TATA oligonucleotide competition. These results demonstrate that TFIIA is an evolutionarily conserved general factor critical for activator-regulated transcription.
Sigma-Aldrich
TFIIA-p12 human, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)
Sigma-Aldrich
TFIIA (p12), γ subunit, GST tagged human, recombinant, expressed in E. coli, ≥70% (SDS-PAGE)