Merck
Palladium-catalyzed oxidative allylic C-H silylation.
Organic letters (2011-03-10)
Johanna M Larsson, Tony S N Zhao, Kálmán J Szabó
ABSTRACT
Palladium-catalyzed allylic C-H silylation was performed with use of hexamethyldisilane as the silyl source. These C-H functionalization reactions occur only in the presence of hypervalent iodine reagents or other strong oxidants and proceed with excellent regioselectivity, providing the linear allylic isomer of the allylsilane products. In demonstrating the first oxidative allylic C-H silylation of alkenes, this study marks an important advance for the catalytic C-H functionalization method.