Merck
  • Assessment of bioavailable B vitamin content in food using in vitro digestibility assay and LC-MS SIDA
Assessment of bioavailable B vitamin content in food using in vitro digestibility assay and LC-MS SIDA
Analytical and Bioanalytical Chemistry (2017)
Paalme T, et al.