Merck
  • Nitric oxide and another potent vasodilator are formed from NG-hydroxy-L-arginine by cultured endothelial cells.
Nitric oxide and another potent vasodilator are formed from NG-hydroxy-L-arginine by cultured endothelial cells.
Proceedings of the National Academy of Sciences of the United States of America (1991-12-15)
A Zembowicz, M Hecker, H Macarthur, W C Sessa, J R Vane
ABSTRACT
The hypothesis was investigated that NG-hydroxy-L-arginine (L-HOArg) is an intermediate in the biosynthesis of nitric oxide (.NO) from L-arginine (L-Arg) by the constitutive .NO synthase (NOS) present in endothelial cells (ECs). When infused through a column of bovine aortic ECs on beads, either L-HOArg or D-HOArg (1-10 microM) substantially potentiated relaxations of the bioassay tissues to .NO released from the cells by ADP or bradykinin, and this effect was abolished by coinfusions of NG-nitro-L-arginine (L-NO2Arg) methyl ester (10 microM) or NG-monomethyl-L-arginine (L-MeArg; 30 microM). Both L-HOArg and D-HOArg, irrespective of the presence of ECs, also potentiated relaxations induced by authentic .NO, but not glyceryl trinitrate. This was due to a rapid chemical reaction of either isomer with .NO, resulting in the formation of a potent and more stable vasodilator. When infusions of L-HOArg (3 microM) were consequently made in the presence of D-HOArg (10 microM), the L-isomer no longer had any effect on relaxations induced by authentic .NO, but significantly increased the stimulated release of .NO from the column of ECs. The conclusion that L-HOArg is a substrate for the constitutive NOS in cultured ECs was strongly supported by the L-NO2Arg-sensitive conversion of L-HOArg, but not D-HOArg, to .NO by NOS preparations from these cells. Interestingly, cultured ECs produced from L-HOArg (greater than or equal to 3 microM), but not D-HOArg, a stable vasodilator, the effects of which were inhibited by oxyhemoglobin (0.3-3 microM). However, the formation of this substance was not prevented by L-NO2Arg methyl ester (10 microM) or L-MeArg (10-100 microM), suggesting an enzymatic pathway different from NOS.
Sigma-Aldrich
Hydroxyguanidine sulfate salt
Pricing and availability is not currently available.
Sigma-Aldrich
NG-Hydroxy-L-arginine, Monoacetate Salt, Cell permeable. A key intermediate in the biosynthesis of nitric oxide by cNOS. NOHA can be efficiently oxidized to nitric oxide and citrulline by cytochrome P450 system.
Pricing and availability is not currently available.