Merck
  • Imaging inflammation using an activated macrophage probe with Slc18b1 as the activation-selective gating target.
Imaging inflammation using an activated macrophage probe with Slc18b1 as the activation-selective gating target.
Nature communications (2019-03-09)
Sung-Jin Park, Beomsue Kim, Sejong Choi, Sivaraman Balasubramaniam, Sung-Chan Lee, Jung Yeol Lee, Heon Seok Kim, Jun-Young Kim, Jong-Jin Kim, Yong-An Lee, Nam-Young Kang, Jin-Soo Kim, Young-Tae Chang
ABSTRACT
Activated macrophages have the potential to be ideal targets for imaging inflammation. However, probe selectivity over non-activated macrophages and probe delivery to target tissue have been challenging. Here, we report a small molecule probe specific for activated macrophages, called CDg16, and demonstrate its application to visualizing inflammatory atherosclerotic plaques in vivo. Through a systematic transporter screen using a CRISPR activation library, we identify the orphan transporter Slc18b1/SLC18B1 as the gating target of CDg16.