Merck
  • Experimental and theoretical electron density studies in large molecules: NAD+, beta-nicotinamide adenine dinucleotide
Experimental and theoretical electron density studies in large molecules: NAD+, beta-nicotinamide adenine dinucleotide
The Journal of Physical Chemistry B (2003)
Guillot B, et al.