Merck
  • Cyclin-dependent Kinase 18 Promotes Oligodendrocyte Precursor Cell Differentiation through Activating the Extracellular Signal-Regulated Kinase Signaling Pathway.
Cyclin-dependent Kinase 18 Promotes Oligodendrocyte Precursor Cell Differentiation through Activating the Extracellular Signal-Regulated Kinase Signaling Pathway.
Neuroscience bulletin (2019-04-28)
Yuchen Pan, Zeping Jiang, Dingya Sun, Zhenghao Li, Yingyan Pu, Dan Wang, Aijun Huang, Cheng He, Li Cao
ABSTRACT
The correct differentiation of oligodendrocyte precursor cells (OPCs) is essential for the myelination and remyelination processes in the central nervous system. Determining the regulatory mechanism is fundamental to the treatment of demyelinating diseases. By analyzing the RNA sequencing data of different neural cells, we found that cyclin-dependent kinase 18 (CDK18) is exclusively expressed in oligodendrocytes. In vivo studies showed that the expression level of CDK18 gradually increased along with myelin formation during development and in the remyelination phase in a lysophosphatidylcholine-induced demyelination model, and was distinctively highly expressed in oligodendrocytes. In vitro overexpression and interference experiments revealed that CDK18 directly promotes the differentiation of OPCs, without affecting their proliferation or apoptosis. Mechanistically, CDK18 activated the RAS/mitogen-activated protein kinase kinase 1/extracellular signal-regulated kinase pathway, thus promoting OPC differentiation. The results of the present study suggest that CDK18 is a promising cell-type specific target to treat demyelinating disease.
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-Olig1 Antibody, ascites fluid, Chemicon®
Sigma-Aldrich
Anti-Myelin Basic Protein Antibody, a.a. 129-138, clone 1, culture supernatant, clone 1, Chemicon®
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid
Sigma-Aldrich
Anti-APC Antibody, clone CC-1, clone CC-1, from mouse
Sigma-Aldrich
Anti-Caspase 3 Antibody, active (cleaved) form, Chemicon®, from rabbit