Merck
  • Modulation of Target Antigen Density Improves CAR T-cell Functionality and Persistence.
Modulation of Target Antigen Density Improves CAR T-cell Functionality and Persistence.
Clinical cancer research : an official journal of the American Association for Cancer Research (2019-05-22)
Sneha Ramakrishna, Steven L Highfill, Zachary Walsh, Sang M Nguyen, Haiyan Lei, Jack F Shern, Haiying Qin, Ira L Kraft, Maryalice Stetler-Stevenson, Constance M Yuan, Jennifer D Hwang, Yang Feng, Zhongyu Zhu, Dimiter Dimitrov, Nirali N Shah, Terry J Fry
ABSTRACT
Chimeric antigen receptor T-cell (CART) therapy targeting CD22 induces remission in 70% of patients with relapsed/refractory acute lymphoblastic leukemia (ALL). However, the majority of post-CD22 CART remissions are short and associated with reduction in CD22 expression. We evaluate the implications of low antigen density on the activity of CD22 CART and propose mechanisms to overcome antigen escape. Using ALL cell lines with variable CD22 expression, we evaluate the cytokine profile, cytotoxicity, and in vivo CART functionality in the setting of low CD22 expression. We develop a high-affinity CD22 chimeric antigen receptor (CAR) as an approach to improve CAR sensitivity. We also assess Bryostatin1, a therapeutically relevant agent, to upregulate CD22 and improve CAR functionality. We demonstrate that low CD22 expression negatively impacts in vitro and in vivo CD22 CART functionality and impairs in vivo CART persistence. Moreover, low antigen expression on leukemic cells increases naïve phenotype of persisting CART. Increasing CAR affinity does not improve response to low-antigen leukemia. Bryostatin1 upregulates CD22 on leukemia and lymphoma cell lines for 1 week following single-dose exposure, and improves CART functionality and in vivo persistence. While Bryostatin1 attenuates IFNγ production by CART, overall in vitro and in vivo CART cytotoxicity is not adversely affected. Finally, administration of Bryostain1 with CD22 CAR results in longer duration of in vivo response. We demonstrate that target antigen modulation is a promising strategy to improve CD22 CAR efficacy and remission durability in patients with leukemia and lymphoma.See related commentary by Guedan and Delgado, p. 5188.
Sigma-Aldrich
Mouse Granzyme B ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Bovine IL2 / Interleukin-2 ELISA Kit, for serum, plasma and cell culture supernatants