Merck
  • Production of generic monoclonal antibody and development of chemiluminescence immunoassay for determination of 32 sulfonamides in chicken muscle.
Production of generic monoclonal antibody and development of chemiluminescence immunoassay for determination of 32 sulfonamides in chicken muscle.
Food chemistry (2019-12-22)
Zhao Bin Li, Peng Lei Cui, Jing Liu, Ju Xiang Liu, Jian Ping Wang
ABSTRACT
A hapten of sulfabenzamide was first synthesized to generate a monoclonal antibody that simultaneously recognized 32 sulfonamides. The computational simulation showed that the 3D conformation, molecular bend angle, molecular volume, electronic charge of core structure of these drugs all showed influences on the antibody binding. The antibody was combined with a heterologous enzyme-labeled hapten to develop a direct competitive chemiluminescence enzyme linked immunosorbent assay for determination of the 32 sulfonamides in chicken muscle sample. The CRs of the optimized method for these drugs were in the range of 7.3%-1778%, and the IC50 values were in the range of 0.038-11.2 ng/g. The limits of detection for detection of these drugs in chicken were in the range of 0.03-26 ng/g. Their recoveries from the standards fortified blank chicken samples were in the range of 60.8%-97.1%. Therefore, this method could be used as a useful tool for routine screening sulfonamides residues in meat.
Sigma-Aldrich
S-Methylisothiourea hemisulfate salt, 98%
Sigma-Aldrich
4-(Imidazol-1-yl)phenol, 97%