Merck
  • Combining native and 'omics' mass spectrometry to identify endogenous ligands bound to membrane proteins.
Combining native and 'omics' mass spectrometry to identify endogenous ligands bound to membrane proteins.
Nature methods (2020-05-07)
Joseph Gault, Idlir Liko, Michael Landreh, Denis Shutin, Jani Reddy Bolla, Damien Jefferies, Mark Agasid, Hsin-Yung Yen, Marcus J G W Ladds, David P Lane, Syma Khalid, Christopher Mullen, Philip M Remes, Romain Huguet, Graeme McAlister, Michael Goodwin, Rosa Viner, John E P Syka, Carol V Robinson
ABSTRACT
Ligands bound to protein assemblies provide critical information for function, yet are often difficult to capture and define. Here we develop a top-down method, 'nativeomics', unifying 'omics' (lipidomics, proteomics, metabolomics) analysis with native mass spectrometry to identify ligands bound to membrane protein assemblies. By maintaining the link between proteins and ligands, we define the lipidome/metabolome in contact with membrane porins and a mitochondrial translocator to discover potential regulators of protein function.
Sigma-Aldrich
Alcohol Dehydrogenase from Saccharomyces cerevisiae, ≥300 units/mg protein, lyophilized powder (contains buffer salts), Mw 141-151 kDa
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand
Sigma-Aldrich
Myoglobin from equine heart, ≥90% (SDS-PAGE), essentially salt-free, lyophilized powder
Sigma-Aldrich
Concanavalin A from Canavalia ensiformis (Jack bean), Type IV, lyophilized powder
Sigma-Aldrich
Tetraethylene glycol monooctyl ether, liquid, ≥98% (GC)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)