Merck
  • Simultaneous identification of multiple receptors of natural product using an optimized cDNA phage display cloning.
Simultaneous identification of multiple receptors of natural product using an optimized cDNA phage display cloning.
Bioorganic & medicinal chemistry letters (2008-06-24)
Qing-Li He, Hui Jiang, Feng Zhang, Hai-Bao Chen, Gong-Li Tang
ABSTRACT
Simultaneously isolating more than one receptor of natural product remains a challenge to chemical genetics. Using cyclosporine A as an affinity probe and an optimized phage display cloning procedure, not only cyclophilin A, but also cyclophilin B was isolated as the full-length gene clone from a human brain cDNA library. This optimized protocol can be used to select protein targets of chemicals dependent on the binding affinity rather than on the relative abundance in cells.
Sigma-Aldrich
Cyclosporin A, BioReagent, from Tolypocladium inflatum, Molecular Biology, ≥95%
Sigma-Aldrich
Cyclosporin A, 97.0-101.5% (on dried basis)
Sigma-Aldrich
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid
Supelco
Cyclosporin A, VETRANAL®, analytical standard