Merck
  • PEG-immobilization of cardol and soluble polymer-supported synthesis of some cardol-coumarin derivatives: preliminary evaluation of their inhibitory activity on mushroom tyrosinase.
PEG-immobilization of cardol and soluble polymer-supported synthesis of some cardol-coumarin derivatives: preliminary evaluation of their inhibitory activity on mushroom tyrosinase.
Bioorganic & medicinal chemistry letters (2008-12-05)
Graziella Tocco, Antonella Fais, Gabriele Meli, Michela Begala, Gianni Podda, M Benedetta Fadda, Marcella Corda, Orazio A Attanasi, Paolino Filippone, Stefano Berretta
ABSTRACT
In this work, the PEG-immobilization and the liquid phase synthesis of some coumarins derived from cardol are presented. Some preliminary results on their tyrosinase inhibitory activity are also included.
Product Description
Supelco
5-Pentadecylresorcinol, analytical standard
Sigma-Aldrich
Kojic acid
Sigma-Aldrich
4-Hexylresorcinol, 98%
Sigma-Aldrich
Resorcinol, BioXtra, ≥99%
Sigma-Aldrich
Resorcinol, ≥98%, FG
Sigma-Aldrich
Resorcinol, ACS reagent, ≥99.0%
Sigma-Aldrich
Resorcinol, ReagentPlus®, 99%