Merck
  • Amides and benzenoids from Zanthoxylum ailanthoides with inhibitory activity on superoxide generation and elastase release by neutrophils.
Amides and benzenoids from Zanthoxylum ailanthoides with inhibitory activity on superoxide generation and elastase release by neutrophils.
Journal of natural products (2009-01-09)
Jih-Jung Chen, Ching-Yi Chung, Tsong-Long Hwang, Jinn-Fen Chen
ABSTRACT
Five new compounds, ailanthamide (1), N-(4-methoxyphenethyl)-N-methylbenzamide (2), (2E,4E)-N-isobutyl-6-oxohepta-2,4-dienamide (3), 4-(4'-hydroxy-3'-methylbutoxy)benzaldehyde (4), and (E)-methyl 4-[4-(3-hydroxypropyl)phenoxy]-2-methylbut-2-enoate (5), and 17 known compounds have been isolated from the stem bark of Zanthoxylum ailanthoides. The structures were determined through spectroscopic and MS analyses. Compounds 1, 3, xanthyletin, decarine, (+)-episesamin, (-)-hinokinin, and evofolin-B exhibited inhibition (IC(50) < or = 5.34 microg/mL) of superoxide anion generation by human neutrophils in response to formyl-l-methionyl-l-leucyl-l-phenylalanine/cytochalasin B (fMLP/CB). Compounds 1, xanthyletin, decarine, and (+)-episesamin also inhibited fMLP/CB-induced elastase release with IC(50) values < or = 5.53 microg/mL.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride solution, ~0.1 M in ethanol (T)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)