Merck
  • Superimposition of metabolic syndrome magnifies post-stenotic kidney injury in dyslipidemic pigs.
Superimposition of metabolic syndrome magnifies post-stenotic kidney injury in dyslipidemic pigs.
American journal of translational research (2021-09-21)
Turun Song, Yu Zhao, Xiangyang Zhu, Alfonso Eirin, James D Krier, Hui Tang, Kyra L Jordan, Amir Lerman, Lilach O Lerman
ABSTRACT
Dyslipidemia aggravates kidney injury distal to atherosclerotic renal artery stenosis (ARAS). Besides dyslipidemia, metabolic syndrome (MetS) also involves development of obesity and insulin-resistance (IR). We hypothesized that concurrent obesity and IR magnify swine stenotic-kidney damage beyond dyslipidemia. Pigs with unilateral RAS were studied after 16 weeks of atherogenic diets without (ARAS) or with (MetS + RAS) development of obesity/IR (n=6 each). Additional pigs on normal diet served as normal or non-dyslipidemic RAS controls (n=6 each). Stenotic-kidney renal blood flow (RBF), glomerular filtration rate (GFR), and microvascular architecture were studied using CT, and oxygenation was studied using blood oxygen level-dependent magnetic-resonance-imaging. We further compared kidney adiposity, oxidative stress, inflammation, apoptosis, fibrosis, and systemic levels of oxidative and inflammatory cytokines. ARAS and MetS + RAS developed hypertension and dyslipidemia, and MetS + RAS also developed obesity and IR. RBF and GFR were similarly decreased in all post-stenotic pig kidneys compared to normal pig kidneys, yet MetS + RAS aggravated and expanded medullary hypoxia and microvascular loss. RAS and ARAS increased systemic levels of tumor necrosis factor (TNF)-α, which were further elevated in MetS + RAS. Renal oxidative stress and TNF-α expression increased in ARAS and further in MetS + RAS, which also upregulated expression of anti-angiogenic angiostatin, and magnified apoptosis, tubular injury, and fibrosis. Beyond dyslipidemia, obesity and insulin-resistance aggravate damage in the post-stenotic kidney in MetS, despite relative hyperfiltration-related preservation of renal function. These observations underscore the need to control systemic metabolic disturbances in order to curb renal damage in subjects with ischemic kidney disease.
Product Description
Millipore
MILLIPLEX® Porcine Cytokine/Chemokine Panel, Configurable Porcine Cytokine/Chemokine 13-Plex Panel