Merck
  • Classic Type 1 Diabetes Mellitus and Fulminant Type 1 Diabetes Mellitus: Similarity and Discrepancy of Immunological Characteristics and Cytokine Profile.
Classic Type 1 Diabetes Mellitus and Fulminant Type 1 Diabetes Mellitus: Similarity and Discrepancy of Immunological Characteristics and Cytokine Profile.
Diabetes, metabolic syndrome and obesity : targets and therapy (2021-12-09)
Lingwen Ying, Yong Zhang, Jun Yin, Yufei Wang, Wei Lu, Wei Zhu, Yuqian Bao, Jian Zhou
ABSTRACT
We aimed to explore the immunological characteristics and cytokine profile of the initial stage of type 1 diabetes. In total, 123 age- and sex-matched subjects with newly diagnosed classic type 1 diabetes mellitus (T1ADM), fulminant type 1 diabetes mellitus (FT1DM), and normal glucose tolerance (NGT) were enrolled. Serum cytokine levels were measured using Milliplex MAP multifactor detection. There was a significant increase in the levels of transforming growth factor β (TGFβ1) and TGFβ2 and decrease in programmed death-1 (PD-1), PD ligand 1 (PD-L1), pro-inflammatory cytokines, and anti-inflammatory cytokines in type 1 diabetes patients compared with the NGT subjects (all P < 0.05). There was no significant difference in C-reactive protein (CRP) and blood routine indicators between the two groups. Type 1 diabetes was further divided into T1ADM and FT1DM subgroups. FT1DM patients had much higher CRP levels than T1ADM patients (4.90 [0.95-26.05] mg/L vs 0.39 [0.20-0.74] mg/L, P < 0.01). Blood routine results showed that the number of leukocytes was significantly increased in FT1DM compared with that in T1ADM (9.2 [5.1-18.8] × 109 cells/L vs 5.4 [4.5-6.7] × 109 cells/L, P < 0.01). In FT1DM patients, neutrophil% was increased, and lymphocyte% was declined significantly, compared with that in T1ADM patients (neutrophil%: 80.2 [59.2-85.2]% vs 59.5 [54.8-64.0]%; lymphocyte%: 18.3 [10.1-32.3]% vs 32.6 [26.8-35.9]%; both P < 0.01). However, there was no difference between FT1DM and T1ADM in cytokine profile except for the decrease in CTLA-4 in T1ADM (P < 0.05). Compared with T1ADM, CRP and leukocytes' levels were increased significantly in FT1DM, with an increase in neutrophil% and decline in lymphocyte%, suggesting that FT1DM may have more abrupt onset and occur as a more serious subtype of type 1 diabetes mellitus.
Product Description
Millipore
MILLIPLEX® Multi-Species TGFß Panel, Premixed Multi-Species TGFß 3-Plex Panel