Merck
  • The Role of Lipid Rafts and Membrane Androgen Receptors in Androgen's Neurotoxic Effects.
The Role of Lipid Rafts and Membrane Androgen Receptors in Androgen's Neurotoxic Effects.
Journal of the Endocrine Society (2022-03-22)
Oluwadarasimi Fadeyibi, Nataliya Rybalchenko, Steve Mabry, Dianna H Nguyen, Rebecca L Cunningham
ABSTRACT
Sex differences have been observed in multiple oxidative stress-associated neurodegenerative diseases. Androgens, such as testosterone, can exacerbate oxidative stress through a membrane androgen receptor (mAR), AR45, localized to lipid rafts in the plasma membrane. The goal of this study is to determine if interfering with mAR localization to cholesterol-rich lipid rafts decreases androgen induced neurotoxicity under oxidative stress environments. We hypothesize that cholesterol-rich caveolar lipid rafts are necessary for androgens to induce oxidative stress generation in neurons via the mAR localized within the plasma membrane. Nystatin was used to sequester cholesterol and thus decrease cholesterol-rich caveolar lipid rafts in a neuronal cell line (N27 cells). Nystatin was applied prior to testosterone exposure in oxidatively stressed N27 cells. Cell viability, endocytosis, and protein analysis of oxidative stress, apoptosis, and mAR localization were conducted. Our results show that the loss of lipid rafts via cholesterol sequestering blocked androgen-induced oxidative stress in cells by decreasing the localization of mAR to caveolar lipid rafts.
Sigma-Aldrich
N27 Rat Dopaminergic Neural Cell Line, The N27 rat dopaminergic neural cell line has been used as a dopaminergic neuron model for in vitro and in vivo studies.
Sigma-Aldrich
Anti-Spectrin alpha chain (nonerythroid) Antibody, clone AA6, clone AA6, Chemicon®, from mouse