Merck
  • Targeting the EIF2AK1 signaling pathway rescues red blood cell production in SF3B1-mutant myelodysplastic syndromes with ringed sideroblasts.
Targeting the EIF2AK1 signaling pathway rescues red blood cell production in SF3B1-mutant myelodysplastic syndromes with ringed sideroblasts.
Blood cancer discovery (2022-08-05)
Vera Adema, Feiyang Ma, Rashmi Kanagal-Shamanna, Natthakan Thongon, Guillermo Montalban-Bravo, Hui Yang, Scott A Peslak, Feng Wang, Pamela Acha, Francesc Sole, Pamela Lockyer, Margherita Cassari, Jaroslaw P Maciejewski, Valeria Visconte, Irene Ganan-Gomez, Yuanbin Song, Carlos Bueso-Ramos, Matteo Pellegrini, Tuyet M Tan, Rafael Bejar, Jennifer S Carew, Stephanie Halene, Valeria Santini, Gheath Al-Atrash, Karen Clise-Dwyer, Guillermo Garcia-Manero, Gerd A Blobel, Simona Colla
ABSTRACT
SF3B1 mutations, which occur in 20% of patients with myelodysplastic syndromes (MDS), are the hallmarks of a specific MDS subtype, MDS with ringed sideroblasts (MDS-RS), which is characterized by the accumulation of erythroid precursors in the bone marrow and primarily affects the elderly population. Here, using single-cell technologies and functional validation studies of primary SF3B1-mutant MDS-RS samples, we show that SF3B1 mutations lead to the activation of the EIF2AK1 pathway in response to heme deficiency and that targeting this pathway rescues aberrant erythroid differentiation and enables the red blood cell maturation of MDS-RS erythroblasts. These data support the development of EIF2AK1 inhibitors to overcome transfusion dependency in patients with SF3B1-mutant MDS-RS with impaired red blood cell production.
Sigma-Aldrich
holo-Transferrin human, ≥98%
Sigma-Aldrich
Insulin solution human, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
5-Azacytidine, ≥98% (HPLC)
Sigma-Aldrich
Human Serum, from human male AB plasma, USA origin, sterile-filtered