Merck
  • Novel human pluripotent stem cell-derived hypothalamus organoids demonstrate cellular diversity.
Novel human pluripotent stem cell-derived hypothalamus organoids demonstrate cellular diversity.
iScience (2023-08-30)
Lily Sarrafha, Drew R Neavin, Gustavo M Parfitt, Ilya A Kruglikov, Kristen Whitney, Ricardo Reyes, Elena Coccia, Tatyana Kareva, Camille Goldman, Regine Tipon, Gist Croft, John F Crary, Joseph E Powell, Joel Blanchard, Tim Ahfeldt
ABSTRACT
The hypothalamus is a region of the brain that plays an important role in regulating body functions and behaviors. There is a growing interest in human pluripotent stem cells (hPSCs) for modeling diseases that affect the hypothalamus. Here, we established an hPSC-derived hypothalamus organoid differentiation protocol to model the cellular diversity of this brain region. Using an hPSC line with a tyrosine hydroxylase (TH)-TdTomato reporter for dopaminergic neurons (DNs) and other TH-expressing cells, we interrogated DN-specific pathways and functions in electrophysiologically active hypothalamus organoids. Single-cell RNA sequencing (scRNA-seq) revealed diverse neuronal and non-neuronal cell types in mature hypothalamus organoids. We identified several molecularly distinct hypothalamic DN subtypes that demonstrated different developmental maturities. Our in vitro 3D hypothalamus differentiation protocol can be used to study the development of this critical brain structure and can be applied to disease modeling to generate novel therapeutic approaches for disorders centered around the hypothalamus.
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, clone LNC1, ascites fluid, clone LNC1, Chemicon®
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, ascites fluid, clone GA5, Chemicon®
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in a glass vial
Sigma-Aldrich
Anti-Dermo-1 Antibody, from rabbit, purified by affinity chromatography