Merck
  • Isolation of a complementary DNA that encodes the mammalian splicing factor SC35.
Isolation of a complementary DNA that encodes the mammalian splicing factor SC35.
Science (New York, N.Y.) (1992-04-24)
X D Fu, T Maniatis
ABSTRACT
The mammalian splicing factor SC35 is required for the first step in the splicing reaction and for spliceosome assembly. The cloning and characterization of a complementary DNA encoding this protein revealed that it is a member of a family of splicing factors that includes mammalian SF2/ASF. This family of proteins is characterized by the presence of a ribonucleoprotein (RNP)-type RNA binding motif and a carboxyl-terminal serine-arginine-rich (SR) domain. A search of the DNA sequence database revealed that the thymus-specific exon (ET) of the c-myb proto-oncogene is encoded on the antisense strand of the SC35 gene.
Sigma-Aldrich
SC35 human, recombinant, expressed in insect cells, ≥80% (SDS-PAGE)
Pricing and availability is not currently available.