Merck
  • A homozygous missense mutation (A659D) in the myophosphorylase gene in a Spanish patient with McArdle's disease.
A homozygous missense mutation (A659D) in the myophosphorylase gene in a Spanish patient with McArdle's disease.
Neuromuscular disorders : NMD (2000-07-19)
M A Martín, J C Rubio, Y Campos, J R Ricoy, A Cabello, J Arenas
ABSTRACT
We identified a novel missense mutation in the myophosphorylase gene (PYGM) in a Spanish patient with McArdle's disease. This homozygous C-to-A mutation results in the replacement of a highly conserved alanine at amino acid position 659 with an aspartic acid in the C-terminal domain of the myophosphorylase gene protein, near binding sites for pyridoxal phosphate and glucose. Our data further expand the genetic heterogeneity in patients with McArdle's disease.
Sigma-Aldrich
Phosphorylase b from rabbit muscle, For use as a marker in SDS-PAGE
Sigma-Aldrich
Phosphorylase b from rabbit muscle, lyophilized powder, ≥20 units/mg protein, 2× crystallization
Sigma-Aldrich
Phosphorylase a from rabbit muscle, lyophilized powder, 20-30 units/mg protein