Merck
  • Molecular cloning of the cDNA for the catalytic subunit of human DNA polymerase epsilon.
Molecular cloning of the cDNA for the catalytic subunit of human DNA polymerase epsilon.
The Journal of biological chemistry (1993-05-15)
T Kesti, H Frantti, J E Syväoja
ABSTRACT
The cDNA encoding the catalytic polypeptide of human DNA polymerase epsilon was cloned. The deduced amino acid sequence reveals that the catalytic polypeptide is 2257 amino acids in length and its calculated molecular mass is 258 kDa. A single RNA message of 7.5 kilobases was recognized by isolated cDNA clones. The identity of the cDNA was verified by direct amino acid sequencing of tryptic fragments derived from the catalytic polypeptide of the HeLa DNA polymerase epsilon. The primary structure comparison with multiple DNA polymerases indicates that human DNA polymerase epsilon catalytic polypeptide is a homolog of the yeast Saccharomyces cerevisiae DNA polymerase II catalytic polypeptide. The proteins are 39% identical. In the region containing known DNA polymerase consensus motifs, the identity is 63%. The expression of the mRNA encoding DNA polymerase epsilon is strongly dependent on cell proliferation.
Sigma-Aldrich
DNA Polymerase I from Escherichia coli lysogenic for NM 964, buffered aqueous glycerol solution
Sigma-Aldrich
Taq DNA Polymerase from Thermus aquaticus, with 10× PCR reaction buffer without MgCl2
Sigma-Aldrich
Taq DNA Polymerase from Thermus aquaticus, with 10× PCR reaction buffer containing MgCl2