Merck
  • Genetic dissection of gustatory sensitivity to bitterness (sucrose octaacetate) in mice.
Genetic dissection of gustatory sensitivity to bitterness (sucrose octaacetate) in mice.
Comptes rendus de l'Academie des sciences. Serie III, Sciences de la vie (1999-12-28)
I Le Roy, J Pager, P L Roubertoux
ABSTRACT
Genes implicated in consumption of a bitter compound, sucrose octaacetate (SOA), were investigated using a full genomic scanning strategy. For a 0.1 mM concentration, two QTL reached 5.8 and 6.5 lod scores on chromosomes 2 (77 cM) and 11 (14 cM), respectively. For a 1 mM concentration, the Soa linkage on chromosome 6 (58 cM, lod score 9.4) was replicated, and another QTL was found on chromosome 19 (15 cM, lod score 3.2). Candidacy of previously identified genes in the close vicinity of the peak of the QTL was examined.
Sigma-Aldrich
Sucrose octaacetate, ≥97%, FG
Sigma-Aldrich
D-(+)-Sucrose octaacetate, 98%