Merck
  • Lipophilicity of a series of 1,2-benzisothiazol-3(2H)-ones determined by reversed-phase thin-layer chromatography.
Lipophilicity of a series of 1,2-benzisothiazol-3(2H)-ones determined by reversed-phase thin-layer chromatography.
Journal of chromatography. A (2002-06-18)
Tomasz Sławik, Cezary Kowalski
ABSTRACT
The lipophilicity (R(Mo)) and specific hydrophobic surface area of seven 1,2-benzisothiazol-3(2H)-ones have been determined by reversed-phase TLC and the effect of different mobile-phase modifiers (acetone, acetonitrile, methanol) on the retention has been studied. The linear correlations between the volume fraction of the organic solvent and the R(M) values over a limited range were established for each solute with high values of correlation coefficients (>0.99). The influence of solvent pH on R(M) values was investigated.
Sigma-Aldrich
1,2-Benzisothiazol-3(2H)-one, 97%
Supelco
1,2-Benzisothiazol-3(2H)-one, analytical standard