Merck
  • EPR and 1H and 13C dynamic nuclear polarization studies of a molecularly doped polymer: bisphenol A polycarbonate doped with trianisylamine and trianisylaminium perchlorate.
EPR and 1H and 13C dynamic nuclear polarization studies of a molecularly doped polymer: bisphenol A polycarbonate doped with trianisylamine and trianisylaminium perchlorate.
Solid state nuclear magnetic resonance (1992-06-01)
R A Wind, N Zumbulyadis, R H Young, Y Hung, L Li, R H Nuttall, G Maciel
ABSTRACT
We have investigated the EPR and DNP behavior of a molecularly doped polymer modeling those used to transport electronic charge in electrophotography. The EPR spectra show no evidence of the superexchange reported for a closely related system based on tri-p-tolylamine. The difference may be due to larger charge-transfer matrix elements in the latter system. An unambiguous interpretation of the observed 1H DNP was rendered difficult by the unanticipated asymmetry of the EPR spectra. We report extensive data on the "three-spin" effect evident in the DNP-enhanced 13C NMR spectra, and comment on its potential for characterizing polymer interfaces.
Product Description
Supelco
Poly(Bisphenol A carbonate), analytical standard
Sigma-Aldrich
Poly(Bisphenol A carbonate), average Mw ~45,000 by GPC
Supelco
Poly(Bisphenol A carbonate)
Sigma-Aldrich
Poly(Bisphenol A carbonate), melt index 10-12 g/10 min (300°C/1.2kg)